Anti-DOT1L Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB64077-100EA 11700 NOK
ABCAAB64077-100
Anti-DOT1L Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DOT1L (DOT1-like histone H3K79 methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat