Anti-MS4A1 Rabbit Monoclonal Antibody [clone: SP32]

Leverandør: Abcam
ABCAAB64088-500EA 11850 NOK
ABCAAB64088-500
Anti-MS4A1 Rabbit Monoclonal Antibody [clone: SP32]
Antistoffer
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone: SP32
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR