Anti-KRT20 Rabbit Monoclonal Antibody [clone: SP33]

Leverandør: Abcam
ABCAAB64091-7EA 11400 NOK
ABCAAB64091-7
Anti-KRT20 Rabbit Monoclonal Antibody [clone: SP33]
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: monoclonal
Clone: SP33
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR