Anti-CD45R Rat Monoclonal Antibody [clone: RA3-6B2]

Leverandør: Abcam
ABCAAB64100-50EA 9025 NOK
ABCAAB64100-50
Anti-CD45R Rat Monoclonal Antibody [clone: RA3-6B2]
Antistoffer
Type: Primary
Antigen: CD45R (CD45R)
Clonality: monoclonal
Clone: RA3-6B2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR