Anti-MYL9 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64161-50EA 11650 NOK
ABCAAB64161-50
Anti-MYL9 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MYL9 (myosin, light chain 9, regulatory)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR