Anti-OAS3 Rabbit polyclonal antibody unconjugated

ABCAAB64163-50EA 7725 NOK
ABCAAB64163-50
Anti-OAS3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: OAS3 (2'-5'-oligoadenylate synthetase 3, 100kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR