Anti-EXOC2 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB64166-50EA 8050 NOK
ABCAAB64166-50
Anti-EXOC2 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: EXOC2 (exocyst complex component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR