Anti-ARHGEF10 Rabbit polyclonal antibody unconjugated

ABCAAB64170-50EA 7175 NOK
ABCAAB64170-50
Anti-ARHGEF10 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ARHGEF10 (Rho guanine nucleotide exchange factor (GEF) 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR