Anti-CHD2 Rabbit polyclonal antibody unconjugated

ABCAAB64174-50EA 7550 NOK
ABCAAB64174-50
Anti-CHD2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CHD2 (chromodomain helicase DNA binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR