Anti-HSP90B2P Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64182-50EA 10900 NOK
ABCAAB64182-50
Anti-HSP90B2P Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSP90B2P (heat shock protein 90kDa beta (Grp94), member 2, pseudogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR