Anti-Mast Cell Tryptase Rabbit Polyclonal Antibody

ABCAAB64192-100EA 7600 NOK
ABCAAB64192-100
Anti-Mast Cell Tryptase Rabbit Polyclonal Antibody
Type: Primary
Antigen: Mast Cell Tryptase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR