Anti-CARD14 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB64366-50EA 8550 NOK
ABCAAB64366-50
Anti-CARD14 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CARD14 (caspase recruitment domain family, member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR