Anti-Oligodendrocyte Specific Protein Rabbit polyclonal antibody unconjugated

ABCAAB64369-100EA 7425 NOK
ABCAAB64369-100
Anti-Oligodendrocyte Specific Protein Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: Oligodendrocyte Specific Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR