Anti-Homeobox protein extradenticle Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64430-100EA 7950 NOK
ABCAAB64430-100
Anti-Homeobox protein extradenticle Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Homeobox protein extradenticle
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR