Anti-Growth Hormone Sheep Polyclonal Antibody

Leverandør: Abcam
ABCAAB64499-250EA 11475 NOK
ABCAAB64499-250
Anti-Growth Hormone Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Growth Hormone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR