Anti-Propranolol Sheep Polyclonal Antibody

Leverandør: Abcam
ABCAAB64519-500EA 8050 NOK
ABCAAB64519-500
Anti-Propranolol Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Propranolol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR