Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-148]
Leverandør: Abcam
ABCAAB64604-50EA 5575 NOK
ABCAAB64604-50
Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-148]
Antistoffer
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone: MEM-148
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human