Anti-CCR5 Mouse monoclonal antibody unconjugated [clone: mAbcam 64620]

ABCAAB64620-100EA 7600 NOK
ABCAAB64620-100
Anti-CCR5 Mouse monoclonal antibody unconjugated [clone: mAbcam 64620]
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: monoclonal
Clone: mAbcam 64620
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR