Anti-C12orf5 Mouse Monoclonal Antibody [clone: M2 - P4H2]
Leverandør: Abcam
ABCAAB64622-100EA 12200 NOK
ABCAAB64622-100
Anti-C12orf5 Mouse Monoclonal Antibody [clone: M2 - P4H2]
Antistoffer
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: monoclonal
Clone: M2 - P4H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human