Anti-C1q Rabbit polyclonal antibody unconjugated

ABCAAB64632-100EA 7600 NOK
ABCAAB64632-100
Anti-C1q Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: C1q
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR