Anti-IgG Goat Polyclonal Antibody (Biotin)
Leverandør: Abcam
ABCAAB64666-500EA 10500 NOK
ABCAAB64666-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå