Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-267]
Leverandør: Abcam
ABCAAB64676-100EA 11400 NOK
ABCAAB64676-100
Anti-HLA DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-267]
Antistoffer
Type: Primary
Antigen: HLA DR
Clonality: monoclonal
Clone: MEM-267
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human