Anti-KITLG Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64677-100EA 12175 NOK
ABCAAB64677-100
Anti-KITLG Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KITLG (KIT ligand)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR