Anti-MPZ Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64685-100EA 8500 NOK
ABCAAB64685-100
Anti-MPZ Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MPZ (myelin protein zero)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR