Anti-APBB2 Rabbit polyclonal antibody unconjugated

ABCAAB64749-50EA 7175 NOK
ABCAAB64749-50
Anti-APBB2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: APBB2 (amyloid beta (A4) precursor protein-binding, family B, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR