Anti-TP53I3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64798-100EA 11675 NOK
ABCAAB64798-100
Anti-TP53I3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TP53I3 (tumor protein p53 inducible protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR