Anti-CYP21A2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64887-100EA 11650 NOK
ABCAAB64887-100
Anti-CYP21A2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP21A2 (cytochrome P450, family 21, subfamily A, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR