Anti-ST5 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64897-50EA 8050 NOK
ABCAAB64897-50
Anti-ST5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ST5 (suppression of tumorigenicity 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR