Anti-IF3EI Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64900-100EA 10900 NOK
ABCAAB64900-100
Anti-IF3EI Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IF3EI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR