Anti-FAM129A Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB64903-50EA 7950 NOK
ABCAAB64903-50
Anti-FAM129A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FAM129A (family with sequence similarity 129, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR