Anti-FCN2 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB64960-100EA 11475 NOK
ABCAAB64960-100
Anti-FCN2 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FCN2 (ficolin (collagen/fibrinogen domain containing lectin) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR