Anti-NR2F6 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65012-100EA 11775 NOK
ABCAAB65012-100
Anti-NR2F6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NR2F6 (nuclear receptor subfamily 2, group F, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat