Anti-FTH1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65080-100EA 12125 NOK
ABCAAB65080-100
Anti-FTH1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR