Anti-HCRTR2 Goat polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB65093-200EA 8425 NOK
ABCAAB65093-200
Anti-HCRTR2 Goat polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: HCRTR2 (hypocretin (orexin) receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR