Anti-CHD7 Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB65097-200EA 8350 NOK
ABCAAB65097-200
Anti-CHD7 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR