Anti-NAA15 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65107-100EA 11650 NOK
ABCAAB65107-100
Anti-NAA15 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NAA15 (N(alpha)-acetyltransferase 15, NatA auxiliary subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human