Anti-TNK2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65108-100EA 11425 NOK
ABCAAB65108-100
Anti-TNK2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TNK2 (tyrosine kinase, non-receptor, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat