Anti-FXR2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65122-100EA 8700 NOK
ABCAAB65122-100
Anti-FXR2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR