Anti-SERPINB5 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65136-100EA 11650 NOK
ABCAAB65136-100
Anti-SERPINB5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SERPINB5 (serpin peptidase inhibitor, clade B (ovalbumin), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR