Anti-ABCB7 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65149-50EA 9975 NOK
ABCAAB65149-50
Anti-ABCB7 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ABCB7 (ATP-binding cassette, sub-family B (MDR/TAP), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR