Anti-HSD3B1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65156-100EA 11475 NOK
ABCAAB65156-100
Anti-HSD3B1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse