Anti-SCN3A Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65164-100EA 11400 NOK
ABCAAB65164-100
Anti-SCN3A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SCN3A (sodium channel, voltage-gated, type III, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human