Anti-Nicotinic Acetylcholine Receptor epsilon Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65180-200EA 8350 NOK
ABCAAB65180-200
Anti-Nicotinic Acetylcholine Receptor epsilon Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor epsilon
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR