Anti-MAPK8IP2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65211-100EA 10875 NOK
ABCAAB65211-100
Anti-MAPK8IP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAPK8IP2 (mitogen-activated protein kinase 8 interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR