Anti-SERINC3 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65218-100EA 11650 NOK
ABCAAB65218-100
Anti-SERINC3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SERINC3 (serine incorporator 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human