Anti-Hepatitis B Virus pre S2 Antigen Mouse Monoclonal Antibody [clone: S9]

Leverandør: Abcam
ABCAAB65406-100EA 8350 NOK
ABCAAB65406-100
Anti-Hepatitis B Virus pre S2 Antigen Mouse Monoclonal Antibody [clone: S9]
Antistoffer
Type: Primary
Antigen: Hepatitis B Virus pre S2 Antigen
Clonality: monoclonal
Clone: S9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR