Anti-AT4G02630 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB65569-50EA 7950 NOK
ABCAAB65569-50
Anti-AT4G02630 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: AT4G02630
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR