Anti-BRAF Rabbit polyclonal antibody unconjugated

ABCAAB65601-400EA 7600 NOK
ABCAAB65601-400
Anti-BRAF Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR