Anti-GSK3B Sheep Polyclonal Antibody
Leverandør: Abcam
ABCAAB65740-100EA 11475 NOK
ABCAAB65740-100
Anti-GSK3B Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse