Anti-STAT5A Sheep Polyclonal Antibody

Leverandør: Abcam
ABCAAB65742-100EA 8500 NOK
ABCAAB65742-100
Anti-STAT5A Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR